5-Bromo-2-cyanopyrimidine, 95%
5-Bromo-2-cyanopyrimidine, 95%
5-Bromo-2-cyanopyrimidine, 95%
5-Bromo-2-cyanopyrimidine, 95%
Thermo Scientific Chemicals

5-Bromo-2-cyanopyrimidine, 95%

CAS: 38275-57-9 | C5H2BrN3 | 183.996 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61363.03
also known as H61363-03
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.10
Save 6.45 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.10
Save 6.45 (14%)
Each
Add to cart
Chemical Identifiers
CAS38275-57-9
IUPAC Name5-bromopyrimidine-2-carbonitrile
Molecular FormulaC5H2BrN3
InChI KeyVPQICCOHFSGBMA-UHFFFAOYSA-N
SMILESBrC1=CN=C(N=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (HPLC)>94.0%
FormSolid
5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-cyanopyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture.
RUO – Research Use Only

General References:

  1. Z. Buděšínský; and J. Vavřina .Nucleophilic substitutions in the 2-methanesulfonylpyrimidine series. Collect. Czech. Chem. Commun. 1972, 37,1721-1733.