4-Amino-5-bromo-2-chloropyrimidine, 95%
4-Amino-5-bromo-2-chloropyrimidine, 95%
4-Amino-5-bromo-2-chloropyrimidine, 95%
Thermo Scientific Chemicals

4-Amino-5-bromo-2-chloropyrimidine, 95%

CAS: 205672-25-9 | C4H3BrClN3 | 208.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61429.06
also known as H61429-06
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS205672-25-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only