2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%
2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%
2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%
2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%
Thermo Scientific Chemicals

2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%

CAS: 1218790-84-1 | C8H5BF3NO2 | 214.938 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H61451.MD
also known as H61451-MD
Price (USD)
159.00
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
159.00
Each
Add to cart
Specifications
CAS1218790-84-1
Chemical Name or Material2-Cyano-4-(trifluoromethyl)benzeneboronic acid
Formula Weight214.94
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only