5-Chlorothiophene-2-carbonyl chloride, 98%
5-Chlorothiophene-2-carbonyl chloride, 98%
5-Chlorothiophene-2-carbonyl chloride, 98%
Thermo Scientific Chemicals

5-Chlorothiophene-2-carbonyl chloride, 98%

CAS: 42518-98-9 | C5H2Cl2OS | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61474.06
also known as H61474-06
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS42518-98-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed container. Store away form water/moisture.
RUO – Research Use Only

General References:

  1. Uroš Trstenjak; Janez Ilaš; and Danijel Kikelj. Spoerri. Transformation of a selective factor Xa inhibitor rivaroxaban into a dual factor Xa/thrombin inhibitor by modification of the morpholin-3-one moiety. Med. Chem. Commun. 2014, 5197-202.
  2. Li, Chao; Liu, Yingshuai; Zhang, Yongjun; Zhang, Xingxian. Spoerri. An approach to the anticoagulant agent rivaroxaban via an isocyanate-oxirane cycloaddition promoted by MgI2.etherate. Journal of Chemical Research. 2011, 35400-401(2).