3-Thiophenemethylamine, 96%
3-Thiophenemethylamine, 96%
3-Thiophenemethylamine, 96%
Thermo Scientific Chemicals

3-Thiophenemethylamine, 96%

CAS: 27757-86-4 | C5H7NS | 113.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61487.03
also known as H61487-03
Price (USD)
164.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
164.00
Each
Chemical Identifiers
CAS27757-86-4
IUPAC Name1-(thiophen-3-yl)methanamine
Molecular FormulaC5H7NS
InChI KeyDUDAKCCDHRNMDJ-UHFFFAOYSA-N
SMILESNCC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Assay from Suppliers CofA≥95.0%
FormLiquid
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store in cool, dry place in tightly closed container. With good ventilation. Store away from air.
RUO – Research Use Only