1-Bromo-2-fluoro-4-nitrobenzene, 98%
1-Bromo-2-fluoro-4-nitrobenzene, 98%
1-Bromo-2-fluoro-4-nitrobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-2-fluoro-4-nitrobenzene, 98%

CAS: 185331-69-5 | C6H3BrFNO2 | 220.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61494.22
also known as H61494-22
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Chemical Identifiers
CAS185331-69-5
IUPAC Name1-bromo-2-fluoro-4-nitrobenzene
Molecular FormulaC6H3BrFNO2
InChI KeyAZYQMHQOHNGPRR-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(F)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only