Methyl 5-amino-2-methylbenzoate, 97%
Methyl 5-amino-2-methylbenzoate, 97%
Methyl 5-amino-2-methylbenzoate, 97%
Thermo Scientific Chemicals

Methyl 5-amino-2-methylbenzoate, 97%

CAS: 18595-12-5 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61671.14
also known as H61671-14
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Chemical Identifiers
CAS18595-12-5
IUPAC Namemethyl 5-amino-2-methylbenzoate
Molecular FormulaC9H11NO2
InChI KeyJNPZKGOLYSCSEL-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)C=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only