5-Bromo-2-fluoroaniline, 98%
5-Bromo-2-fluoroaniline, 98%
5-Bromo-2-fluoroaniline, 98%
5-Bromo-2-fluoroaniline, 98%
Thermo Scientific Chemicals

5-Bromo-2-fluoroaniline, 98%

CAS: 2924-09-6 | C6H5BrFN | 190.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61678.14
also known as H61678-14
Price (USD)
73.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
73.00
Each
Add to cart
Chemical Identifiers
CAS2924-09-6
IUPAC Name5-bromo-2-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyADWKOCXRCRSMLQ-UHFFFAOYSA-N
SMILESNC1=C(F)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
FormPowder or crystalline powder or fused solid or crystalline solid
Assay (GC)≥97.5%
Proton NMRConforms to structure
5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Alfred Courtin. Zur synthese sulfonierter derivate von 2-fluoranilin. Helvetica Chimica Acta. 1983, 66,(1), 68-75.