2-Bromo-3-fluoroaniline, 97%
2-Bromo-3-fluoroaniline, 97%
2-Bromo-3-fluoroaniline, 97%
Thermo Scientific Chemicals

2-Bromo-3-fluoroaniline, 97%

CAS: 111721-75-6 | C6H5BrFN | 190.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61687.03
also known as H61687-03
Price (USD)
93.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
93.80
Each
Chemical Identifiers
CAS111721-75-6
IUPAC Name2-bromo-3-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyXZRSXRUYZXBTGD-UHFFFAOYSA-N
SMILESNC1=C(Br)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Veronica Guilarte; M.Pilar Castroviejo; Patricia Garcia-Garcia; Manuel A.Fernandez-Rodriguez; Roberto Sanz. Approaches to the synthesis of 2,3-dihaloanilines. Useful precursors of 4-functionalized-1H-indoles. J. Org. Chem. 2011, 76,(9), 3416-3437.
  2. T.D.Bradshaw; A.D.Westwell. The development of the antitumour benzothiazole prodrug, phortress, as a clinical candidate. Current Medicinal Chemistry. 2004, 11,(8), 1009-1021.