Potassium thiobenzoate, 95%
Potassium thiobenzoate, 95%
Potassium thiobenzoate, 95%
Potassium thiobenzoate, 95%
Thermo Scientific Chemicals

Potassium thiobenzoate, 95%

CAS: 28170-13-0 | C7H6KOS | 177.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H61741.MD
also known as H61741-MD
Price (USD)
62.65
Online Exclusive
69.40
Save 6.75 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.40
Save 6.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS28170-13-0
IUPAC Namebenzenecarbothioic S-acid potassium
Molecular FormulaC7H6KOS
InChI KeyLSBMSRUYNGEWKI-UHFFFAOYSA-N
SMILES[K].SC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormPowder or crystalline powder
Assay (HPLC)≥94%
Proton NMRConforms to structure
Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiobenzoate is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mary Bean; Harold Kohn. Studies on the reaction of mitomycin C with potassium thiobenzoate under reductive conditions. J. Org. Chem. 1985, 50,(3), 293-298.
  2. J.M.Heap; L.N.Owen. Thio-sugars. Part V. Rates of the nucleophilic substitution reactions of some 1,2:5,6-di-O-isopropylidene-3-O-tosyl-α-D-hexoses with potassium thiobenzoate. J. Chem. Soc. C. 1970, (5), 712-714.