5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals
5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals
5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61748.06
also known as H61748-06
5 g
Catalog number H61748.06
also known as H61748-06
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS20532-33-6
IUPAC Name5-chloro-1-benzothiophene
Molecular FormulaC8H5ClS
InChI KeySNYURIHMNFPQFL-UHFFFAOYSA-N
SMILESClC1=CC=C2SC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Lance S.Fuller; Brian Iddon; Kevin A.Smith. Thienothiophenes. Part 2.1 Synthesis, metallation and bromine→lithium exchange reactions of thieno[3,2-b]thiophene and its polybromo derivatives. J. Chem. Soc., Perkin Trans. 1. 1997, (22), 3465-3470.
  2. S.Perez-Silanes; J.Martinez-Esparza; A.M.Oficialdegui; H.Villanueva; L.Oruas; A.Monge. Synthesis of new 5-substitutedbenzo[b]thiophene derivatives. Journal of Heterocyclic Chemistry. 2001, 38,(5), 1025-1030.