Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%
Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%
Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%
Thermo Scientific Chemicals

Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%

CAS: 4815-32-1 | C8H11NO2S | 185.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61766.06
also known as H61766-06
Price (USD)
401.65
Online Exclusive
446.00
Save 44.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
401.65
Online Exclusive
446.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS4815-32-1
IUPAC Nameethyl 2-amino-5-methylthiophene-3-carboxylate
Molecular FormulaC8H11NO2S
InChI KeyAYBUNZGJQVYGTM-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(N)SC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only