4-(Methoxymethyl)benzeneboronic acid, 97%
4-(Methoxymethyl)benzeneboronic acid, 97%
4-(Methoxymethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

4-(Methoxymethyl)benzeneboronic acid, 97%

CAS: 279262-11-2 | C8H11BO3 | 165.983 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61795.06
also known as H61795-06
Price (USD)
46.70
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.70
Each
Specifications
CAS279262-11-2
Chemical Name or Material4-(Methoxymethyl)benzeneboronic acid
Formula Weight165.99
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only