2-Bromo-3,4-difluoroaniline, 97%
2-Bromo-3,4-difluoroaniline, 97%
2-Bromo-3,4-difluoroaniline, 97%
Thermo Scientific Chemicals

2-Bromo-3,4-difluoroaniline, 97%

CAS: 1092349-87-5 | C6H4BrF2N | 208.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61876.14
also known as H61876-14
Price (USD)
620.65
Online Exclusive
689.00
Save 68.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
620.65
Online Exclusive
689.00
Save 68.35 (10%)
Each
Chemical Identifiers
CAS1092349-87-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White-like to yellowish
FormCrystalline powder
Assay (unspecified)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only