4-Fluoro-2-methoxyaniline, 95%
4-Fluoro-2-methoxyaniline, 95%
4-Fluoro-2-methoxyaniline, 95%
Thermo Scientific Chemicals

4-Fluoro-2-methoxyaniline, 95%

CAS: 450-91-9 | C7H8FNO | 141.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61965.06
also known as H61965-06
Price (USD)
643.65
Online Exclusive
715.00
Save 71.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
643.65
Online Exclusive
715.00
Save 71.35 (10%)
Each
Chemical Identifiers
CAS450-91-9
IUPAC Name4-fluoro-2-methoxyaniline
Molecular FormulaC7H8FNO
InChI KeyBNRRMRUVYDETQC-UHFFFAOYSA-N
SMILESCOC1=C(N)C=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only