N-Boc-D-aspartic acid 4-benzyl ester, 98%
N-Boc-D-aspartic acid 4-benzyl ester, 98%
N-Boc-D-aspartic acid 4-benzyl ester, 98%
Thermo Scientific Chemicals

N-Boc-D-aspartic acid 4-benzyl ester, 98%

CAS: 51186-58-4 | C16H21NO6 | 323.345 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62000.14
also known as H62000-14
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS51186-58-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Bergman, Y.; Ciampini, M.; Jalal, S.; Lagiakos, H. R.; Aguilar, M. I.; Perlmutter, P. The synthesis of Fmoc-O-allyl beta-serine. Tetrahedron: Asymmetry 2008, 19 (24), 2861-2863.
  2. Jungheim, L. N.; Shepherd, T. A.; Baxter, A. J.; Burgess, J.; Hatch, S. D.; Lubbehusen, P.; Wiskerchen, M. A.; Muesing, M. A. Potent Human Immunodeficiency Virus Type 1 Protease Inhibitors That Utilize Noncoded D-Amino Acids as P2/P3 Ligands. J. Med. Chem. 1996, 39 (1), 96-108.