2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%
2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%
2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 1831130-33-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62033.03
also known as H62033-03
Price (USD)
200.65
Online Exclusive
223.00
Save 22.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
200.65
Online Exclusive
223.00
Save 22.35 (10%)
Each
Specifications
CAS1831130-33-6
Chemical Name or Material2-Amino-3-fluorobenzeneboronic acid pinacol ester
Formula Weight237.08
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only