4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals
4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals
4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62040.06
also known as H62040-06
5 g
Catalog number H62040.06
also known as H62040-06
Price (USD)
485.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
485.00
Each
Specifications
CAS304445-49-6
Chemical Name or Material4-Bromo-3-fluoroacetophenone
Formula Weight217.04
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only