4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62059.03
also known as H62059-03
1 g
Catalog number H62059.03
also known as H62059-03
Price (USD)
274.65
Online Exclusive
305.00
Save 30.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
274.65
Online Exclusive
305.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS1288978-82-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
FormPowder
Assay (HPLC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only