(S)-2,3-Diaminopropionic acid monohydrochloride, 95%
(S)-2,3-Diaminopropionic acid monohydrochloride, 95%
(S)-2,3-Diaminopropionic acid monohydrochloride, 95%
Thermo Scientific Chemicals

(S)-2,3-Diaminopropionic acid monohydrochloride, 95%

CAS: 1482-97-9 | C3H9ClN2O2 | 140.567 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62128.06
also known as H62128-06
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1482-97-9
IUPAC Name2,3-diaminopropanoic acid hydrochloride
Molecular FormulaC3H9ClN2O2
InChI KeySKWCZPYWFRTSDD-UHFFFAOYNA-N
SMILESCl.NCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only