2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals
2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals
2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62211.14
also known as H62211-14
25 g
Catalog number H62211.14
also known as H62211-14
Price (USD)
426.65
Special offer
Online exclusive
Ends: 30-Jun-2026
502.00
Save 75.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
426.65
Special offer
Online exclusive
Ends: 30-Jun-2026
502.00
Save 75.35 (15%)
Each
Chemical Identifiers
CAS94744-50-0
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
Molecular FormulaC19H19NO4
InChI KeyHOZZVEPRYYCBTO-UHFFFAOYSA-N
SMILESCC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only