3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62234.03
also known as H62234-03
1 g
Catalog number H62234.03
also known as H62234-03
Price (USD)
568.65
Online Exclusive
632.00
Save 63.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
568.65
Online Exclusive
632.00
Save 63.35 (10%)
Each
Specifications
CAS1887126-77-3
Chemical Name or Material3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester
Formula Weight238.06
IUPAC Name3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
InChI KeyKBGOBMOAZQOCTA-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only