N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%
N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%
N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%

CAS: 4779-31-1 | C19H19NO6 | 357.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62306.06
also known as H62306-06
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS4779-31-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only