trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

CAS: 181308-57-6 | C12H21NO3 | 227.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62338.03
also known as H62338-03
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Add to cart
Specifications
CAS181308-57-6
Chemical Name or Materialtrans-4-(Boc-amino)cyclohexanecarboxaldehyde
Formula Weight227.3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only