Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62378.06
also known as H62378-06
5 g
Catalog number H62378.06
also known as H62378-06
Price (USD)
128.65
Special offer
Online exclusive
Ends: 30-Jun-2026
151.00
Save 22.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
128.65
Special offer
Online exclusive
Ends: 30-Jun-2026
151.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS144978-12-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only