(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%
Thermo Scientific Chemicals

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

CAS: 198544-42-2 | C23H26N2O6 | 426.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62381.06
also known as H62381-06
Price (USD)
493.65
Special offer
Online exclusive
Ends: 30-Jun-2026
581.00
Save 87.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
493.65
Special offer
Online exclusive
Ends: 30-Jun-2026
581.00
Save 87.35 (15%)
Each
Chemical Identifiers
CAS198544-42-2
IUPAC Name3-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC23H26N2O6
InChI KeyPKAUMAVONPSDRW-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only