trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%
trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%
trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%
Thermo Scientific Chemicals

trans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride, 97%

CAS: 1222709-28-5 | C14H21ClN2O2 | 284.784 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62409.03
also known as H62409-03
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
301.00
Save 45.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
301.00
Save 45.35
Each
Specifications
CAS1222709-28-5
Chemical Name or Materialtrans-3-(Benzyloxycarbonylamino)cyclohexylamine hydrochloride
Formula Weight284.78
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only