trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%
trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%
Thermo Scientific Chemicals

trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%

CAS: 149423-77-8 | C14H20N2O2 | 248.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62444.03
also known as H62444-03
Price (USD)
67.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
67.10
Each
Specifications
CAS149423-77-8
Chemical Name or Materialtrans-4-(Benzyloxycarbonylamino)cyclohexylamine
Formula Weight248.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only