(S)-4-Amino-2-(Boc-amino)butyric acid, 97%
(S)-4-Amino-2-(Boc-amino)butyric acid, 97%
(S)-4-Amino-2-(Boc-amino)butyric acid, 97%
Thermo Scientific Chemicals

(S)-4-Amino-2-(Boc-amino)butyric acid, 97%

CAS: 25691-37-6 | C9H18N2O4 | 218.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62450.06
also known as H62450-06
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 30-Jun-2026
198.00
Save 29.35 (15%)
Each
Chemical Identifiers
CAS25691-37-6
IUPAC Name(2S)-4-azaniumyl-2-{[(tert-butoxy)carbonyl]amino}butanoate
Molecular FormulaC9H18N2O4
InChI KeyMDCPCLPRWLKUIQ-LURJTMIESA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC[NH3+])C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only