trans-4-Cyanocyclohexylamine hydrochloride, 97%
trans-4-Cyanocyclohexylamine hydrochloride, 97%
trans-4-Cyanocyclohexylamine hydrochloride, 97%
Thermo Scientific Chemicals

trans-4-Cyanocyclohexylamine hydrochloride, 97%

CAS: 873537-33-8 | C7H13ClN2 | 160.645 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62522.MD
also known as H62522-MD
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS873537-33-8
IUPAC Name4-aminocyclohexane-1-carbonitrile hydrochloride
Molecular FormulaC7H13ClN2
InChI KeyBTLVSLNZSMIPCW-UHFFFAOYSA-N
SMILESCl.NC1CCC(CC1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)>97%
Proton NMRConforms to structure
Carbon-13 NMRConforms to structure
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in a cool and dry condition. Incompatible with oxidizing agents.
RUO – Research Use Only