3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%

CAS: 1073354-74-1 | C17H25BFNO3 | 321.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62530.03
also known as H62530-03
Price (USD)
202.65
Online Exclusive
225.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
202.65
Online Exclusive
225.00
Save 22.35 (10%)
Each
Add to cart
Specifications
CAS1073354-74-1
Chemical Name or Material3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Formula Weight321.19
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only