2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%
2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%
2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 863578-24-9 | C12H17BFNO2 | 237.081 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62575.MD
also known as H62575-MD
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Specifications
CAS863578-24-9
Chemical Name or Material2-Amino-5-fluorobenzeneboronic acid pinacol ester
Formula Weight237.08
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only