cis-4-Isopropylcyclohexanecarboxylic acid, 97%
cis-4-Isopropylcyclohexanecarboxylic acid, 97%
cis-4-Isopropylcyclohexanecarboxylic acid, 97%
cis-4-Isopropylcyclohexanecarboxylic acid, 97%
Thermo Scientific Chemicals

cis-4-Isopropylcyclohexanecarboxylic acid, 97%

CAS: 7084-93-7 | C10H18O2 | 170.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H62594.MD
also known as H62594-MD
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.70
Save 12.05 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.70
Save 12.05 (15%)
Each
Add to cart
Specifications
CAS7084-93-7
Chemical Name or Materialcis-4-Isopropylcyclohexanecarboxylic acid
Formula Weight170.24
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only