3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62652.03
also known as H62652-03
1 g
Catalog number H62652.03
also known as H62652-03
Price (USD)
321.65
Special offer
Online exclusive
Ends: 30-Jun-2026
378.00
Save 56.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
321.65
Special offer
Online exclusive
Ends: 30-Jun-2026
378.00
Save 56.35 (15%)
Each
Specifications
CAS710348-94-0
Chemical Name or Material3-Fluoro-5-nitrobenzeneboronic acid pinacol ester
Formula Weight267.06
IUPAC Name2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyCKBJFSWEKLWHTI-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only