trans-4-(Boc-amino)cyclohexylamine, 97%
trans-4-(Boc-amino)cyclohexylamine, 97%
trans-4-(Boc-amino)cyclohexylamine, 97%
trans-4-(Boc-amino)cyclohexylamine, 97%
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexylamine, 97%

CAS: 177906-48-8 | C11H22N2O2 | 214.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62734.14
also known as H62734-14
Price (USD)
440.65
Online Exclusive
489.00
Save 48.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
440.65
Online Exclusive
489.00
Save 48.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS177906-48-8
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0%
Appearance (Color)White to pale cream
Proton NMRConforms to structure
FormPowder
trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Arbuckle, W., et al. Optimisation of pharmacokinetic properties to afford an orally bioavailable and selective V1A receptor antagonist. J. Chem. Soc., Perkin Trans. 1. 2011, 21 (15), 4622-4628.