N-Boc-N-methyl-L-valine, 95%
N-Boc-N-methyl-L-valine, 95%
N-Boc-N-methyl-L-valine, 95%
N-Boc-N-methyl-L-valine, 95%
Thermo Scientific Chemicals

N-Boc-N-methyl-L-valine, 95%

CAS: 45170-31-8 | C11H21NO4 | 231.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62771.06
also known as H62771-06
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS45170-31-8
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Molecular FormulaC11H21NO4
InChI KeyXPUAXAVJMJDPDH-QMMMGPOBSA-N
SMILESCC(C)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only