(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%
Thermo Scientific Chemicals

(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%

CAS: 3350-13-8 | C29H47N3O6 | 533.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62772.03
also known as H62772-03
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.50
Save 11.85 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.50
Save 11.85 (15%)
Each
Add to cart
Chemical Identifiers
CAS3350-13-8
IUPAC Name2-{[(benzyloxy)carbonyl]amino}-4-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Molecular FormulaC29H47N3O6
InChI KeyCYMIEBREXUYHGN-UHFFFAOYNA-N
SMILESC1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only