3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62811.03
also known as H62811-03
1 g
Catalog number H62811.03
also known as H62811-03
Price (USD)
253.65
Special offer
Online exclusive
Ends: 30-Jun-2026
298.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 30-Jun-2026
298.00
Save 44.35 (15%)
Each
Add to cart
Specifications
CAS445303-67-3
Chemical Name or Material3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester
Formula Weight290.06
IUPAC Name2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyFPCSARGPMNDYAO-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only