Specifications
CAS204245-65-8
Chemical Name or Materialtrans-3-[4-(Boc-amino)cyclohexyl]propionic acid
Formula Weight271.35
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more