N-Boc-S-trityl-D-cysteine, 98%
N-Boc-S-trityl-D-cysteine, 98%
N-Boc-S-trityl-D-cysteine, 98%
Thermo Scientific Chemicals

N-Boc-S-trityl-D-cysteine, 98%

CAS: 87494-13-1 | C27H29NO4S | 463.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62863.06
also known as H62863-06
Price (USD)
240.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
240.00
Each
Chemical Identifiers
CAS87494-13-1
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular FormulaC27H29NO4S
InChI KeyJDTOWOURWBDELG-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only