cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%
Thermo Scientific Chemicals

cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

CAS: 181308-56-5 | C12H21NO3 | 227.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62871.03
also known as H62871-03
Price (USD)
174.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
174.00
Each
Chemical Identifiers
CAS181308-56-5
IUPAC Nametert-butyl N-(4-formylcyclohexyl)carbamate
Molecular FormulaC12H21NO3
InChI KeyGPDBIGSFXXKWQR-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1CCC(CC1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or fused solid or crystalline chunks
Assay from Suppliers CofA≥96.0%
cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only