trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%
trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%
trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%
trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%
Thermo Scientific Chemicals

trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%

CAS: 947141-86-8 | C13H21NO2 | 223.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62905.03
also known as H62905-03
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
401.00
Save 60.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
401.00
Save 60.35 (15%)
Each
Add to cart
Specifications
CAS947141-86-8
Chemical Name or Materialtrans-1-(Boc-amino)-4-ethynylcyclohexane
Formula Weight223.31
IUPAC Nametert-butyl N-(4-ethynylcyclohexyl)carbamate
InChI KeyNZTBBJYAWNVCOE-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only