6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%
6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%
6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%
6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%
Thermo Scientific Chemicals

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%

CAS: 120-15-0 | C10H13NO | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63002.06
also known as H63002-06
Price (USD)
294.65
Special offer
Online exclusive
Ends: 30-Jun-2026
347.00
Save 52.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
294.65
Special offer
Online exclusive
Ends: 30-Jun-2026
347.00
Save 52.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS120-15-0
IUPAC Name6-methoxy-1,2,3,4-tetrahydroquinoline
Molecular FormulaC10H13NO
InChI KeyFRXSZNDVFUDTIR-UHFFFAOYSA-N
SMILESCOC1=CC=C2NCCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Assay from Supplier's CofA≥96.0%
FormSolid
Mass SpecConforms
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Neeraj Shakya; Kuldeep K. Roy; Anil K. Saxena. Substituted 1,2,3,4-tetrahydroquinolin-6-yloxypropanes as β3-adrenergic receptor agonists: Design, synthesis, biological evaluation and pharmacophore modeling. Bioorganic & Medicinal Chemistry. 2009, 17 (2) 830-847.
  2. Tomihiro Nishiyama; Tatsuya Suzuki; Yasuhiro Hashiguchi; Shingo Shiotsu; Masataka Fujioka. Antioxidant activity of aromatic cyclic amine derivatives. Polymer Degradation and Stability. 2002, 75 (3), 549-554.