2-Fluoro-6-nitrobenzoic acid, 98%
2-Fluoro-6-nitrobenzoic acid, 98%
2-Fluoro-6-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

2-Fluoro-6-nitrobenzoic acid, 98%

CAS: 385-02-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H63013.06
also known as H63013-06
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.20
Save 8.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.20
Save 8.55 (15%)
Each
Chemical Identifiers
CAS385-02-4
IUPAC Name2-fluoro-6-nitrobenzoic acid
Molecular FormulaC7H4FNO4
InChI KeyMPDZCNPDHUUPRL-UHFFFAOYSA-N
SMILESOC(=O)C1=C(C=CC=C1F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only