3-(1-Methyl-4-piperidinyl)indole, 97%
3-(1-Methyl-4-piperidinyl)indole, 97%
3-(1-Methyl-4-piperidinyl)indole, 97%
Thermo Scientific Chemicals

3-(1-Methyl-4-piperidinyl)indole, 97%

CAS: 17403-07-5 | C14H18N2 | 214.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Catalog number H63020.06
also known as H63020-06
Price (USD)
826.65
Special offer
Online exclusive
Ends: 28-Dec-2026
972.00
Save 145.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
826.65
Special offer
Online exclusive
Ends: 28-Dec-2026
972.00
Save 145.35
Each
Specifications
Beilstein1573327
CAS17403-07-5
Chemical Name or Material3-(1-Methyl-4-piperidinyl)indole
ColorWhite to Yellow
Formula Weight214.31
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only