This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H63081.22
also known as H63081-22
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Specifications
Beilstein508247
CAS156-41-2
Chemical Name or Material2-(4-Chlorophenyl)ethylamine
DOT InformationHazard Class: 8; Packaging Group: III
Density1.120 g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.