6-Amino-5-bromoquinoline, 97%
6-Amino-5-bromoquinoline, 97%
6-Amino-5-bromoquinoline, 97%
Thermo Scientific Chemicals

6-Amino-5-bromoquinoline, 97%

CAS: 50358-42-4 | C9H7BrN2 | 223.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63150.03
also known as H63150-03
Price (USD)
194.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
194.00
Each
Specifications
CAS50358-42-4
Chemical Name or Material6-Amino-5-bromoquinoline
ColorYellow to Brown
Formula Weight223.07
Health Hazard 1H315-H319-H335
View more
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only