3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%
3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%
3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%
3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%
Thermo Scientific Chemicals

3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%

CAS: 590371-38-3 | C7H4F3NO2 | 191.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63174.06
also known as H63174-06
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS590371-38-3
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only