D-Pipecolinic acid, 97%
D-Pipecolinic acid, 97%
D-Pipecolinic acid, 97%
Thermo Scientific Chemicals

D-Pipecolinic acid, 97%

CAS: 1723-00-8 | C6H11NO2 | 129.159 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H63255.22
also known as H63255-22
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
401.00
Save 60.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
401.00
Save 60.35 (15%)
Each
Chemical Identifiers
CAS1723-00-8
IUPAC Namepiperidine-2-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyHXEACLLIILLPRG-UHFFFAOYNA-N
SMILESOC(=O)C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only