L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%
L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%

CAS: 2673-19-0 | C9H18ClNO4 | 239.696 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63257.06
also known as H63257-06
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2673-19-0
IUPAC Name4-tert-butyl 1-methyl 2-aminobutanedioate hydrochloride
Molecular FormulaC9H18ClNO4
InChI KeySFYKWYAIJZEDNG-UHFFFAOYNA-N
SMILESCl.COC(=O)C(N)CC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only